Beskrivning
Equipped with our exclusive anti-fog and anti-scratch PLATINUM coating on both sides of the lens, our range of autoclavable multi-use goggles are resistant to high temperature and still fully functional after several autoclaving cycles. Our goggles’ designs fit over any prescription glasses, ensuring improved eye protection and vision. Overflow chute for liquids Upper ventilation / sealed in the lower part Compatible with a half-mask respirator PLATINUM coating : anti-scratch & anti-fog
Specifikationer
Specifikationer
| Linstyp | Platinum |
| Produkttyp | Goggles |
| Ram färg | Translucent |
| Disponibel | Reusable |
| Linsmaterial | Polycarbonate |
| Rammaterial | TPR |
| Ramtyp | Direct ventilated on the upper part and sealed on the bottom |
| Sterilitet | Sterile |