Läs mer
Cytiva Superdex™ 75 3.2/300 Prepacked Precision Column
Handla allt Cytiva Produkter
Flödeshastighet:
0.01 to 0.10 mL/min.
Förpackningsstorlek:
Varje
Beskrivning
- Prepacked with Superdex 75 media
- For molecules with molecular weight between 3,000 and 70,000
- Spherical composite of cross-linked agarose and dextran
- Supplied with two fingertight 1/16 in. male connectors for connection
- For polishing step in purification process
- With new hardware design for direct connection to ÄATKAM systems
Specifikationer
Specifikationer
| Flödeshastighet | 0.01 to 0.10 mL/min. |
| Produkttyp | Superdex Pre-packed Column |
| Material | Borosilicate Glass |