Learn More
Systec™ Superdry for H-Series Autoclaves
Systec™ Superdry for H-Series Autoclaves increases the drying efficiency for solids and porous materials such as filters and fabrics.
Brand:  Systec™ 9608
Additional Details : Weight : 1.00000kg
Product Code. 12512777
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Heat energy from the standard steam generator is transferred to the heating coils around the body of the sterilization chamber and is used for drying.
  • Deep-vacuum drying using the optional vacuum device in conjunction with Superdry avoids the necessity for subsequent drying in a separate drying cabinet.
Specifications
Specifications
Increasing autoclave drying efficiency
H-Series Autoclaves
Superdry Option for Autoclave