Learn More
Systec™ Superdry for H-Series Autoclaves
Systec™ Superdry for H-Series Autoclaves increases the drying efficiency for solids and porous materials such as filters and fabrics.
Brand: Systec™ 9608
Additional Details : Weight : 1.00000kg
Description
- Heat energy from the standard steam generator is transferred to the heating coils around the body of the sterilization chamber and is used for drying.
- Deep-vacuum drying using the optional vacuum device in conjunction with Superdry avoids the necessity for subsequent drying in a separate drying cabinet.
Specifications
Increasing autoclave drying efficiency | |
H-Series Autoclaves | |
Superdry Option for Autoclave |