missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperFrost™ Microscope Slides, Ground 45°

The printed Thermo Scientific™ SuperFrost tab is impervious to virtually all common laboratory chemicals and reagents. A wide range of colors (7 standard colors) allows several examination specimens to be labeled and archived simply and clearly.

Brand:  Thermo Scientific™ ABAA000085##32E00MNZ10MH#

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15233069

  • 845.00 SEK / Pack of 1000


Estimated Shipment
19-09-2018
Add to basket

Description

Description

  • Time-consuming labeling with paper labels is no longer required
  • The writing area is resistant to all common solvents
  • SuperFrost slides may be sorted by color e.g. by patient, urgency, stain or examination method
  • Simple recognition of specified characteristics such as year of examination or department, use etc.
Specifications

Specifications

76mm
1mm
ISO8037/I
Ground 45°
Yes
26mm
Orange
Uncoated
50/Box, 5,000 outer carton
76 x 26mm
Videos