missing translation for 'onlineSavingsMsg'
Learn More

SUPT3H Recombinant Protein, Abnova

Brand:  Abnova H00008464-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16132482

  • 3343.96 SEK / 10µg

Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH50384
Solution
8464
SUPT3H (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SUPT3H
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
61.82
8
Glutathione Sepharose 4 Fast Flow
10 ug
MVVFGTTFFNFDSFIGKTDSIEIIGKSVFPYCYHNILPLLNDSGRYSLGDARRPLHETAVLVEDVVHTQLINLLQQAAEVSQLRGARVITPEDLLFLMRKDKKKLRRLLKYMFIRDYKSKIVKGIDEDDLLEDKLSGSNNANKRQKIAQDFLNSIDQTGELLAMFEDDEIDEVKQERMERAERQTRIMDSAQYAEFCESRQLSFSKKASKFRDWLDCSSMEIKPNVVAMEILAYLAYETVAQLVDLALLVRQDMVTKAGDPFSHAISATFIQYHNSAESTAACGVEAHSDAIQPCHIREAIRRYSHRIGPLSPFTNAYRRNGMAFLAC
SPT3/SPT3L
SUPT3H
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates