missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUPT7L Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | SUPT7L |
---|---|
Concentration | 0.39 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16824175
|
Proteintech
25606-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16814175
|
Proteintech
25606-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
SUPT7L is a protein subunit of the human STAGA complex (SPT3)/TAF9/GCN5 acetyltransferase complex), which is a chromatin-modifying multiprotein complex.Specifications
SUPT7L | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O94864 | |
9913 | |
SUPT7L Fusion Protein Ag22264 | |
Primary | |
-20°C | |
SUPT7L |
0.39 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Adenocarcinoma antigen ART1, ART1, KIAA0764, SPT7L, STAF65, STAF65(gamma), STAF65gamma, STAGA complex 65 subunit gamma, Suppressor of Ty 7 like, SUPT7L | |
SUPT7L | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |