Learn More
Invitrogen™ SureCast™ Multi-Use Tool, 10-well
These components of the SureCast Handcast System can be used to create polyacrylamide gels using SureCast Handcast Reagents as well as any other popular polyacrylamide gel casting reagents.
Brand: Invitrogen™ HC1010
Additional Details : Weight : 0.13500kg
Description
- Pour polyacrylamide gels easily and confidently
- For use with the SureCast vertical gel system
- SureCast gels are designed for use in the Mini Gel Tank
Recommended Storage
Room Temperature
Services
Up to 12 months
Specifications
SureCast Vertical Gel System | |
SureCast | |
Room Temperature |
Mini | |
SureCast™ Multi-use Tool, 10-well |
For Research Use Only. Not for use in diagnostic procedures.