Läs mer
Surfactin, TRC
Produktkod. 30011273
Change view
Kvantitet:
1 mg
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30011273 1 mg -
30007815 2.5 mg 2.5mg
30005376 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011273 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS8600031MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 24730-31-2
Molekylformel C53H93N7O13
Molekylvikt (g/mol) 1036.34
Synonym Surfactin C1, 1-Oxa-4,7,10,13,16,19,22-heptaazacyclopentacosane, cyclic peptide deriv., iso-C15-Surfactin, N-[N-[N-[N-[N-[N-[N-(3-Hydroxy-13-methyltetradecanoyl)-L-α-glutamyl]-L-leucyl]-D-leucyl]-L-valyl]-L-α-aspartyl]-D-leucyl]-L-leucine ψ-Lactone, Cyclo(L-α-aspartyl-D-leucyl-L-leucyl-3-hydroxy-13-methyltetradecanoyl-L-α-glutamyl-L-leucyl-D-leucyl-L-valyl)
LEDER O[13C@]1[13C@](O)[13C@@](O)[13C@](O[C@H]2[C@H](O)[C@@H](O)C(O)O[C@@H]2CO)O[13C@@]1[13C]O
Specifikationer
Kemiskt namn eller material Surfactin
Kvantitet 1 mg
InChI formel InChI=1S/C53H93N7O13/c1-30(2)20-18-16-14-13-15-17-19-21-36-28-43(61)54-37(22-23-44(62)63)47(66)55-38(24-31(3)4)48(67)57-40(26-33(7)8)51(70)60-46(35(11)12)52(71)58-41(29-45(64)65)50(69)56-39(25-32(5)6)49(68)59-42(27-34(9)10)53(72)73-36/h30-42,46H,13-29H2,1-12H3,(H,54,61)(H,55,66)(H,56,69)(H,57,67)(H,58,71)(H,59,68)(H,60,70)(H,62,63)(H,64,65)/t36-,37+,38+,39-,40-,41+,42+,46+/m1/s1
Formel vikt 1035.6831