missing translation for 'onlineSavingsMsg'
Learn More

Survivin Ab-1, Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Survivin Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-1201-P0

Additional Details : Weight : 0.00170kg

 View more versions of this product

Product Code. 12639256

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Research Areas: Apoptosis; Survivin, a new inhibitor of apoptosis (IAP) protein, is expressed in the G2/M phase of the cell cycle in a cycle-regulated manner. At the beginning of mitosis, survivin associates with microtubules of the mitotic spindle in a specific and saturable reaction that is regulated by microtubule dynamics. Disruption of survivin-microtubule interactions results in loss of survivin's anti-apoptosis function and increased caspase-3 activity, a mechanism involved in cell death, during mitosis. It indicates that survivin may counteract a default induction of apoptosis in G2/M phase. The overexpression of survivin in cancer may overcome this apoptotic checkpoint and favour aberrant progression of transformed cells through mitosis.

Host Species: Mouse

Clone: 8E2

Isotype: IgG1/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: aa 1-99

Immunogen: Full-length recombinant human survivin protein

Molecular Weight: 16.5kDa

Positive Control: HeLa cells

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting (Not suitable)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Survivin Ab-1
Monoclonal
200μg/mL
Mouse
100 μL
Apoptosis
Human, Rat
IgG2a κ
Immunofluorescence
8E2
Unconjugated
Full length recombinant human survivin protein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers