Learn More
Survivin Ab-1, Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Survivin Ab-1, Mouse Monoclonal Antibody.
1183.20 SEK - 5778.30 SEK
Specifications
Antigen | Survivin Ab-1 |
---|---|
Clone | 8E2 |
Applications | Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
12649256
|
Epredia
MS-1201-P1 |
200μg/mL | 500 μL |
4845.00 SEK
500µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12639256
|
Epredia
MS-1201-P0 |
200μg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12629256
|
Epredia
MS-1201-P |
200μg/mL | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12659256
|
Epredia
MS-1201-P1ABX |
1mg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12679256
|
Epredia
MS-1201-PABX |
1mg/mL | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Research Areas: Apoptosis; Survivin, a new inhibitor of apoptosis (IAP) protein, is expressed in the G2/M phase of the cell cycle in a cycle-regulated manner. At the beginning of mitosis, survivin associates with microtubules of the mitotic spindle in a specific and saturable reaction that is regulated by microtubule dynamics. Disruption of survivin-microtubule interactions results in loss of survivin's anti-apoptosis function and increased caspase-3 activity, a mechanism involved in cell death, during mitosis. It indicates that survivin may counteract a default induction of apoptosis in G2/M phase. The overexpression of survivin in cancer may overcome this apoptotic checkpoint and favour aberrant progression of transformed cells through mitosis.Host Species: Mouse
Clone: 8E2
Isotype: IgG1/κ
Species Reactivity: Human and Rat. Others not known.
Epitope: aa 1-99
Immunogen: Full-length recombinant human survivin protein
Molecular Weight: 16.5kDa
Positive Control: HeLa cells
Cellular Localization: Cytoplasmic and nuclear
Recommended for:
- Immunohistochemistry (Not suitable)
- Western Blotting (Not suitable)
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
Survivin Ab-1 | |
Immunofluorescence | |
Unconjugated | |
Mouse | |
Apoptosis | |
Full length recombinant human survivin protein | |
Primary |
8E2 | |
Monoclonal | |
Concentrated | |
RUO | |
Human, Rat | |
IgG2a κ |