Learn More
SV40 Large T Antigen Ab-1 Mouse Monoclonal Antibody, Epredia™
Epredia™ SV40 Large T Antigen Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
2965.00 SEK - 8600.00 SEK
Specifications
Antigen | SV40 Large T Antigen Ab-1 |
---|---|
Clone | PAb101; same as clone 412 |
Applications | Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
12600237
|
Epredia
MS-1832-P0 |
200μg/mL | 100 μL |
2965.00 SEK
100µL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12610237
|
Epredia
MS-1832-P1 |
200μg/mL | 500 μL |
6410.00 SEK
500µL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12690227
|
Epredia
MS-1832-P |
200μg/mL | 1 mL |
8600.00 SEK
1mL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12620237
|
Epredia
MS-1832-P1ABX |
1mg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12630237
|
Epredia
MS-1832-PABX |
1mg/mL | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
The simian virus 40 large T antigen plays important role in SV40 transfection and transformation of the cells. It binds to Rb and p53 suppressor proteins leading to the inactivation of suppressor pathway.Host Species: Mouse
Clone: PAb101; originally clone 412
Isotype: IgG2a1
Species Reactivity: SV40 virus.
Epitope: C-terminal
Immunogen: SV40-transformed mouse line B4
Molecular Weight: 94kDa
Positive Control: SV40-transfected cells
Recommended for:
- Western Blotting
- Immunofluorescence
- Immunoprecipitation
Note:
USA: RUO; Int'l: RUO
Specifications
SV40 Large T Antigen Ab-1 | |
Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Infectious Disease | |
SV40 transformed mouse line B4 |
PAb101; same as clone 412 | |
Monoclonal | |
Concentrated | |
RUO | |
Simian virus 40 (SV40) | |
Primary |