Learn More
SVL™ Coupling for Butt Joint
Plastic coupling and sealing rings designed to secure two SVR™ glass screwthreads of the same dia. In a butt joint
Brand:  SVL 701-14
Additional Details : Weight : 0.05040kg
 View more versions of this product
Product Code. 12003133
Quantity Price
1 223.10 SEK / Each
10 222.94 SEK / Each
In stock, ready to ship within 9 to 11 days: 8
Add to basket
Description
Description
  • Plastic coupling and sealing ring designed to secure two SVL™ glass screwthreads of the same diameter in a butt joint
  • Coupling is manufactured from phenolic plastic and consists of two separately threaded sections
  • Sealing ring is manufactured from silicone rubber and sheathed with PTFE for protection against chemical attack. Internal diameter of the ring corresponds to the bore of the SVL™ glass screwthread so that flow is not restricted
  • Heat resistant up to 200°C
Specifications
Specifications
Plastic
SVL™ and Quickfit™ Glassware
Sealing ring
Joint Size: 42
To secure two SVL™ Glass screw threads of the same diameter in a butt joint