Learn More
Pyrex™ SVL™ Stirrer Heads for SVL™ Stirrer Shafts
Switch stirring heads quickly and easily. Pyrex™ SVL™ Stirrer Head for SVL™ Stirrer Shafts are constructed of durable PTFE polymer; some types are fortified with a metal core.
Brand: Pyrex™ 588-95
1458.25 SEK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.02000kg
Description
- Designed for use with SVL™ stirrer shafts
- Head simply screws into the end of the shaft
- Manufactured from a PTFE based polymer with the Anchor and Helix types stiffened by a metal core
Specifications
90 mm | |
SVL™ reaction vessels | |
1 Each | |
Anchor |