Läs mer
Swainsonine (Synthetic), TRC
Produktkod. 30008294
Change view
Kvantitet:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008294 25 mg
17903055 5 mg
17952845 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008294 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS88500025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 72741-87-8
Molekylformel C8 H15 N O3
Molekylvikt (g/mol) 173.21
Synonym Swainsonine
IUPAC-namn (1S,2R,8R,8aR)-1,2,3,5,6,7,8,8a-octahydroindolizine-1,2,8-triol
LEDER O[C@@H]1CCCN2C[C@@H](O)[C@@H](O)[C@@H]12
Specifikationer
Kemiskt namn eller material Swainsonine
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C8H15NO3/c10-5-2-1-3-9-4-6(11)8(12)7(5)9/h5-8,10-12H,1-4H2/t5-,6-,7-,8-/m1/s1
Formel vikt 173.1052
Procent renhet >95%