Läs mer
(-)-Sweroside, TRC
Produktkod. 30138190
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138190 100 mg
30004370 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138190 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS885530100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14215-86-2
Molekylformel C16 H22 O9
Molekylvikt (g/mol) 358.34
Synonym 1H,3H-Pyrano[3,4-c]pyran-1-one, 5-ethenyl-6-(β-D-glucopyranosyloxy)-4,4a,5,6-tetrahydro-, (4aS,5R,6S)-, 1H,3H-Pyrano[3,4-c]pyran-1-one, 5-ethenyl-6-(β-D-glucopyranosyloxy)-4,4a,5,6-tetrahydro-, [4aS-(4aα,5β,6α)]-, Sweroside (8CI), (-)-Sweroside, (4aS,5R,6S)-5-Ethenyl-6-(β-D-glucopyranosyloxy)-4,4a,5,6-tetrahydro-1H,3H-pyrano[3,4-c]pyran-1-one
IUPAC-namn (3S,4R,4aS)-4-ethenyl-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4,4a,5,6-tetrahydro-3H-pyrano[3,4-c]pyran-8-one
LEDER OC[C@H]1O[C@@H](O[C@@H]2OC=C3[C@@H](CCOC3=O)[C@H]2C=C)[C@H](O)[C@@H](O)[C@@H]1O
Specifikationer
Kemiskt namn eller material Sweroside
Kvantitet 100 mg
InChI formel InChI=1S/C16H22O9/c1-2-7-8-3-4-22-14(21)9(8)6-23-15(7)25-16-13(20)12(19)11(18)10(5-17)24-16/h2,6-8,10-13,15-20H,1,3-5H2/t7-,8+,10-,11-,12+,13-,15+,16+/m1/s1
Formel vikt 358.1264