Learn More
Hettich™ Swing-out Rotor for ROTINA 380/380 R Centrifuge, 4 place
Product Code. 10696083
Change view
Click to view available options
Max. Speed:
4,000 rpm
Unit Size:
Each
Product Code. Max. Speed Unit Size
10696083 4,000 rpm Each
1 options
Product Code. 10696083 Supplier Hettich™ Supplier No. 1798
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Beräknad leverans: 07-01-2027
Provides consistent, accurate results for blood, cell/tissue culture, plates or bottle applications. The ROTINA 380 R is a high capacity, general purpose, benchtop centrifuge with a small footprint.
  • For use with ROTINA 380/380 R
  • The ROTINA 380's versatility and size make an invaluable addition to any lab. Ideal for clinical, research, food and beverage and industrial applications.
  • Robust and steady high-capacity centrifuge
  • Accessories with bio-containment protect users from aerosols while ensuring optimal safety, for instance, while centrifuging infectious samples
  • Temperature control system with pre-cool function from -20 to +40°C
  • Versa-Rotor™ for multiple applications
  • All-steel construction
  • Quiet Operation
  • Easy-Lift™/Safety-Lock™ Lid
Specifications
Type Swing-out Rotor
Max. RCF 3,095 x g
Max. Speed 4,000 rpm
Radius (Metric) 168 mm
Radius (Metric) Min. 142 mm
Max. Tube Length 115 mm
Max. Tube Diameter 65 mm
For Use With (Equipment) ROTINA 380/380 R
Angle 90°
No. of Places 4