Learn More
SYCP2L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557012
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TSMICVIEDF FDTALIISRS SSEGKIQMLD SFLLSLGFLV TEKTVNHLLQ QEGLKTFNCI LHAVPREERK KFPLSEGMCH LMKDLARTLL TVGDYDQQV Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 64%.
SYCP2L is a protein coding gene.Specifications
SYCP2L | |
Polyclonal | |
Unconjugated | |
SYCP2L | |
145 kDa nucleolar protein homolog; C6orf177; dJ62D2.1; hsNO145; NO145; SCP-2-like; SYCP2L; synaptonemal complex protein 2 like; synaptonemal complex protein 2-like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
221711 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T4T6 | |
SYCP2L | |
Recombinant protein corresponding to Human SYCP2L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only