Läs mer
Invitrogen™ Human Syk, GST Tag Recombinant Protein
Beskrivning
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length, Recombinant, Cytoplasmic.
Specifikationer
Specifikationer
| Tillträdesnummer | P43405 |
| Koncentration | See Label |
| För användning med (applikation) | Kinase Assay |
| Formulering | 50mM tris HCl with 0.5mM EDTA, 150mM NaCl, 0.02% Triton X-100, 50% glycerol, 2mM DTT and no preservative; pH 7.5 |
| Gen-ID (Entrez) | 6850 |
| Molekylvikt (g/mol) | 100.1 kDa |
| Namn | Human Syk, GST Tag |
| pH-intervall | 7.5 |
| Reningsmetod | Purified |
| Kvantitet | 10 μg |
| Visa mer |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.