Läs mer
(+)-Sylvatesmin, TRC
Produktkod. 30428227
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30428227 5 mg
1 options
Produktkod. 30428227 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS2130205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (+)-Sylvatesmin
CAS 487-39-8
Molekylformel C21 H24 O6
Kvantitet 5 mg
Synonym 4-[(1S,3aR,4R,6aR)-4-(3,4-Dimethoxyphenyl)tetrahydro-1H,3H-furo[3,4-c]furan-1-yl]-2-methoxyphenol, (+)-Phillygenin, (+)-Phylligenin, Epipinoresinol Methyl Ether, Forsythigenol, Phillygenin, Phillygenol, Phillyrin aglycone, Sylvatesmin
InChI formel InChI=1S/C21H24O6/c1-23-17-7-5-13(9-19(17)25-3)21-15-11-26-20(14(15)10-27-21)12-4-6-16(22)18(8-12)24-2/h4-9,14-15,20-22H,10-11H2,1-3H3/t14-,15-,20+,21-/m0/s1
LEDER COc1ccc(cc1OC)[C@@H]2OC[C@H]3[C@@H]2CO[C@@H]3c4ccc(O)c(OC)c4
IUPAC-namn 4-[(3S,3aR,6R,6aR)-6-(3,4-dimethoxyphenyl)-1,3,3a,4,6,6a-hexahydrofuro[3,4-c]furan-3-yl]-2-methoxyphenol
Molekylvikt (g/mol) 372.412
Formel vikt 372.157