Learn More
Synapsin 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565185
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: IGEHQVEDRQL ITELVISKMN QLLSRTPALS PQRPLTTQQP QSGTLKDPDS SKTPP Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.
Synapsin 2 Polyclonal antibody specifically detects Synapsin 2 in Human samples. It is validated for Immunocytochemistry.Specifications
Synapsin 2 | |
Polyclonal | |
Unconjugated | |
SYN2 | |
2900074L19Rik; AI836018; AI841723; SYN2; synapsin 2; synapsin II; Synapsin-2; SYNII; SYNIIa; SYNIIb | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6854 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q92777 | |
SYN2 | |
Recombinant Human Synapsin 2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only