missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptogyrin 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10432-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Synaptogyrins comprise a family of tyrosine-phosphorylated proteins with two neuronal (synaptogyrins 1 and 3) isoforms. Previous studies have indicated that synaptogyrins are involved in the regulation of neurotransmitter release. Synaptogyrin 1 is an integral membrane protein ubiquitously expressed in the nervous system. Synaptogyrin has four transmembrane domains and an extended cytoplasmic tail but it is not glycosylated. Synaptogyrin 1 is a tyrosine-phosphorylated by c-src and possibly other tyrosine kinases. The function of Synaptogyrin 1 is not well understood, but it may be a useful marker for nerve terminals and diagnosing neuroendocrine tumors.Specifications
Synaptogyrin 1 | |
Polyclonal | |
Unconjugated | |
SYNGR1 | |
synaptogyrin 1, SYNGR1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20972, 29205, 9145 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43759, O55100, Q62876 | |
SYNGR1 | |
Synaptogyrin 1 Fusion Protein Ag0685 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |