missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptotagmin-9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12793-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
SYT9 may be involved in Ca2+-dependent exocytosis of secretory vesicles through Ca2+ and phospholipid binding to the C2 domain or may serve as Ca2+ sensors in the process of vesicular trafficking and exocytosis.Specifications
Synaptotagmin-9 | |
Polyclonal | |
Unconjugated | |
SYT9 | |
Synaptotagmin 9, synaptotagmin IX, SYT9, SytIX | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
143425, 60510, 60564 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q86SS6, Q925C0, Q9R0N9 | |
Syt9 | |
Synaptotagmin-9 Fusion Protein Ag3520 | |
150 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |