missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYNDIG1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27537-1-AP-20UL
Additional Details : Weight : 0.00010kg
Specifications
SYNDIG1 | |
Polyclonal | |
Unconjugated | |
SYNDIG1 | |
C20orf39, Dispanin subfamily C member 2, DSPC2, Synapse differentiation-inducing gene protein 1, SynDIG1, TMEM90B, Transmembrane protein 90B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
433485, 79953 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A2ANU3, Q9H7V2 | |
Syndig1 | |
SYNDIG1 Fusion Protein Ag26442 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |