missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYNDIG1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | SYNDIG1 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16825685
|
Proteintech
27537-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16815685
|
Proteintech
27537-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SYNDIG1 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
A2ANU3, Q9H7V2 | |
433485, 79953 | |
SYNDIG1 Fusion Protein Ag26442 | |
Primary | |
-20°C | |
SYNDIG1 |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C20orf39, Dispanin subfamily C member 2, DSPC2, Synapse differentiation-inducing gene protein 1, SynDIG1, TMEM90B, Transmembrane protein 90B | |
Syndig1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |