Läs mer
Synephrine-13C2,15N Hydrochloride Salt, TRC
Produktkod. 30425246
Change view
Kvantitet:
0.1 mg
1 mg
Förpackningsstorlek:
0.1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30425246 1 mg -
30409711 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30425246 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS9200021MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1329499-30-0
Molekylformel C713C2H1315NO2 • HCl
Molekylvikt (g/mol) 206.65
Synonym 4-Hydroxy-α-[(methylamino)methyl]benzenemethanol-13C2,15N, N-[2-Hydroxy-2-(4-hydroxyphenyl)ethyl]-N-methylamine-13C2,15N, dl-1-(4-Hydroxyphenyl)-2-(methylamino)ethanol-13C2,15N, p-Hydroxy-α-[(methylamino)methyl]benzyl Alcohol-13C2,15N, NSC 166285-13C2,15N, (+/-)-N-Methyloctopamine-13C2,15N, Analeptin-13C2,15N, DL-Synephrine-13C2,15N, Ethaphene-13C2,15N
IUPAC-namn 4-(1-hydroxy-2-(methylamino-15N)ethyl-1,2-13C2)phenol hydrochloride
LEDER OC1=CC=C([13C]([13C]([H])([H])[15N]([H])C)([H])O)C=C1.Cl
Specifikationer
Kemiskt namn eller material Synephrine-13C2,15N Hydrochloride Salt
Kvantitet 1 mg
InChI formel InChI=1S/C9H13NO2.ClH/c1-10-6-9(12)7-2-4-8(11)5-3-7;/h2-5,9-12H,6H2,1H3;1H/i6+1,9+1,10+1;
Formel vikt 206.08