Learn More
SYNM Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560647
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MDVSNVEAIR SRTQEAGALG VSDRGSWRDA DSRNDQAVGV SFKASAGEGD QAHREQGKEQ AMFDKKVQLQ RMVDQRSVIS D Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 56%.
Type-VI intermediate filament (IF) which plays an important cytoskeletal role within the muscle cell cytoskeleton. It forms heteropolymeric IFs with desmin and/or vimentin, and via its interaction with cytoskeletal proteins alpha-dystrobrevin, dystrophin, talin-1, utrophin and vinculin, is able to link these heteropolymeric IFs to adherens-type junctions, such as to the costameres, neuromuscular junctions, and myotendinous junctions within striated muscle cells.Specifications
SYNM | |
Polyclonal | |
Unconjugated | |
SYNM | |
4930412K21Rik; AI852401; Desmuslin; DMN; E130104F11; KIAA0353; SYN; synemin; synemin alpha; synemin beta; synemin, intermediate filament protein; synm | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23336 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15061 | |
SYNM | |
Recombinant protein corresponding to Human SYNM | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only