missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 1A / Syntaxin 1B Mouse anti-Human, Mouse, Rat, Clone: 2D3B5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66437-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the syntaxin superfamily. Syntaxins are nervous system-specific proteins implicated in the docking of synaptic vesicles with the presynaptic plasma membrane. Syntaxins possess a single C-terminal transmembrane domain, a SNARE domain, and an N-terminal regulatory domain. Syntaxins bind synaptotagmin in a calcium-dependent fashion and interact with voltage dependent calcium and potassium channels via the C-terminal H3 domain. This gene product is a key molecule in ion channel regulation and synaptic exocytosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
Syntaxin 1A / Syntaxin 1B | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35526, P32851, P61264, P61265, P61266, Q16623 | |
STX1A, Stx1b | |
Syntaxin 1A / Syntaxin 1B Fusion Protein Ag10257 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
2D3B5 | |
Unconjugated | |
STX1A | |
STX1B, STX1B1, STX1B2, syntaxin 1B, Syntaxin 1B1, Syntaxin 1B2 | |
Mouse | |
Protein G | |
RUO | |
112755, 116470, 20907, 24923, 56216, 6804 | |
-20°C | |
Liquid |