Learn More
Sartorius Syringe Holder/Support for Sterisart™ 16469
The pre-filled Syringe Holder/Support for Sterisart™ 16469 facilitates dilution of the contents of pre-filled syringes and flushing the surface of a syringe needle, during sterility testing.
Brand: Sartorius 1ZA---0024
Additional Details : Weight : 0.86000kg
Description
- Syringe holder and rinsing fluid flow-rate controller/restrictor
- Compatible with:
- Sterisart™ 16469--------GBD
- Sterisart™ 16469--------GSD
Specifications
Sterisart System 16469 | |
Syringe Holder |