Learn More
SYT17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15413-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody works well in WB, IHC and IF applications.
Plays a role in dendrite formation by melanocytes.Specifications
SYT17 | |
Polyclonal | |
Unconjugated | |
SYT17 | |
Protein B/K, Synaptotagmin 17, synaptotagmin XVII, SYT17, SytXVII | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
110058, 192189, 51760 | |
-20°C | |
Liquid |
Immunofluorescence, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q62807, Q920M7, Q9BSW7 | |
SYT17 | |
SYT17 Fusion Protein Ag7691 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |