missing translation for 'onlineSavingsMsg'
Learn More

SYT17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15413-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824574

  • 3313.85 SEK / 150µL
Estimated Shipment: 03-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody works well in WB, IHC and IF applications.

Plays a role in dendrite formation by melanocytes.
Specifications

Specifications

SYT17
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q62807, Q920M7, Q9BSW7
Protein B/K, Synaptotagmin 17, synaptotagmin XVII, SYT17, SytXVII
Rabbit
IgG
150 μL
-20°C
Primary
110058, 192189, 51760
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
SYT17
Liquid
SYT17
SYT17 Fusion Protein Ag7691
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers