Läs mer
Tocris Bioscience™ T 112
Beskrivning
T 112 is an estrogen-related receptor gamma (ERRγ) agonist. Promotes maturation of hiPSC-derived cardiomyocytes. T 112 accelerates metabolic and sarcomere maturation in cardiomyocytes, as well as maturation of contractile and electrophysiological properties and ultrastructure organization, when used alone or in combination with other small molecules.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2-[2-(4-Bromophenyl)-1H-benzimidazol-6-yl]-2,3-dihydro-5-(4-morpholinyl)-1H-isoindol-1-one |
| CAS | 2376876-87-6 |
| Kvantitet | 10 mg |
| Mål | Estrogen and Related Receptor Agonists |
| Molekylformel | C25H21BrN4O2 |