Läs mer
T-705 Ribofuranose, TRC
Produktkod. 30004605
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004605 5 mg
30003959 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30004605 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT0032005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 356782-88-2
Molekylformel C10H12FN3O6
Molekylvikt (g/mol) 289.22
Synonym 6-Fluoro-3,4-dihydro-3-oxo-4-β-D-ribofuranosyl-2-pyrazinecarboxamide, 6-Fluoro-3,4-dihydro-3-oxo-4-β-D-ribofuranosyl-pyrazinecarboxamide;
LEDER O[C@H]([C@H]1CO)[C@@H](O)[C@@H](O1)N2C(C(C(N)=O)=NC(F)=C2)=O
Specifikationer
Kemiskt namn eller material T-705 Ribofuranose
Kvantitet 5 mg
InChI formel InChI=1S/C10H12FN3O6/c11-4-1-14(9(19)5(13-4)8(12)18)10-7(17)6(16)3(2-15)20-10/h1,3,6-7,10,15-17H,2H2,(H2,12,18)/t3-,6-,7-,10-/m1/s1
Formel vikt 289.071