missing translation for 'onlineSavingsMsg'
Läs mer

T-705RTP Sodium Salt (>75%), TRC

Produktkod. 30407146
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30407146 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. View return policy
Product Code. 30407146 Supplier Toronto Research Chemicals Supplier No. TRCF5979251MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material T-705RTP Sodium Salt
CAS 356783-10-3
Molekylformel C10 H15 F N3 O15 P3 . xNa
Kvantitet 1 mg
Synonym 6-Fluoro-3,4-dihydro-4-[5-O-[hydroxy[[hydroxy(phosphonooxy)phosphinyl]oxy]phosphinyl]-β-D-ribofuranosyl]-3-oxo-pyrazinecarboxamide Sodium Salt
InChI formel InChI=1S/C10H15FN3O15P3.Na/c11-4-1-14(9(18)5(13-4)8(12)17)10-7(16)6(15)3(27-10)2-26-31(22,23)29-32(24,25)28-30(19,20)21;/h1,3,6-7,10,15-16H,2H2,(H2,12,17)(H,22,23)(H,24,25)(H2,19,20,21);/q;+1/t3-,6-,7-,10-;/m1./s1
LEDER [Na+].NC(=O)C1=NC(=CN([C@@H]2O[C@H](COP(=O)(O)OP(=O)(O)OP(=O)(O)O)[C@@H](O)[C@H]2O)C1=O)F
IUPAC-namn sodium;[[(2R,3S,4R,5R)-5-(3-carbamoyl-5-fluoro-2-oxo-pyrazin-1-yl)-3,4-dihydroxy-tetrahydrofuran-2-yl]methoxy-hydroxy-phosphoryl] phosphono hydrogen phosphate
Molekylvikt (g/mol) 529.16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.