missing translation for 'onlineSavingsMsg'
Läs mer

t-Boc-aminooxyacetic Acid N-Hydroxysuccinimide Ester, TRC

Produktkod. 30333490
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30333490 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30333490 Supplier Toronto Research Chemicals Supplier No. TRCB200760100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material t-Boc-aminooxyacetic Acid N-Hydroxysuccinimide Ester
CAS 80366-85-4
Molekylformel C11 H16 N2 O7
Kvantitet 100 mg
Synonym 2-[[[(1,1-Dimethylethoxy)carbonyl]amino]oxy]acetic Acid 2,5-Dioxo-1-pyrrolidinyl Ester, [2-[(2,5-Dioxo-1-pyrrolidinyl)oxy]-2-oxoethoxy]carbamic Acid 1,1-Dimethylethyl Ester
InChI formel InChI=1S/C11H16N2O7/c1-11(2,3)19-10(17)12-18-6-9(16)20-13-7(14)4-5-8(13)15/h4-6H2,1-3H3,(H,12,17)
LEDER CC(C)(C)OC(=O)NOCC(=O)ON1C(=O)CCC1=O
IUPAC-namn (2,5-dioxopyrrolidin-1-yl) 2-[(2-methylpropan-2-yl)oxycarbonylamino]oxyacetate
Molekylvikt (g/mol) 288.25
Formel vikt 288.0958
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.