Läs mer
T Cell Receptor Gamma/Delta Hamster, anti-Mouse, Biotin, Clone: GL3, STEMCELL Technologies
Hamster Monoclonal Antibody
Brand:  Stemcell Technologies 60104BT.2
Encompass_Preferred
 Se fler versioner av denna produkt
Produktkod. 17119031
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
  • The GL3 antibody reacts with the δ chain of the murine T cell receptor γ/δ (TCRγ/δ or TCR gamma/delta), a subtype of the TCR involved in the recognition of both peptide and lipid antigens
  • TCRγ/δ comprises a heterodimer (∼ 80 kDa in humans) of disulfide-linked γ and δ subunits that associates with CD3 on the cell surface
  • It is a member of the immunoglobulin superfamily
  • TCRγ/δ is expressed on a subpopulation of T cells in the circulation but may be found on up to 50% of the T cells in epithelial cell-rich tissues
  • These cells have roles in oral and tumor-associated tolerance as well as autoimmune disease and have been described as a link between the adaptive and innate immune responses
  • Once activated, they secrete effector cytokines in a subtype- and context-specific manner
  • Most γ/δ T cells are CD4-/CD8-, though some express CD8
  • The GL3 antibody recognizes an epitope in the constant region of the δ chain and can reportedly activate TCRγ/δ+ cells
Specifikationer
Specifikationer
T Cell Receptor Gamma/Delta
Monoclonal
Biotin
50 μg
Mouse
T Cells, T Cells, Other Subsets
Immunohistochemistry, Flow Cytometry, Immunofluorescence, Functional Assay, Blocking Assay
GL3
Hamster
Primary
IgG2 κ
Produktförslag
Produktförslag
Filmer
SDS
Dokument
Dokument
Certifikat
Kampanjer
Kampanjer
Korrigering av produktinnehåll
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.