Learn More
Masterflex™ T-Connecting Union, PVDF
Cole-Parmer™ T-Connecting Union, PVDF is barbed, designed for quick and easy assembly, economical, and suitable for low-pressure applications.
Brand:  Masterflex™ 30703-79
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12429640
  • 713.00 SEK / Pack of 10
Estimated Shipment: 13-07-2022
to see stock.
Add to basket
Description
Description
  • Ideal for low-pressure applications.
  • Quick and easy assembly without tools.
  • Available in a variety of sizes, and configurations.
  • Easily connect fluid paths using flexible tubing.
Specifications
Specifications
Autoclavable
5/8 in. ID tubing
10/Pk.
Fits 5/8 in. ID tubing
Barb
Low-pressure applications
T-connector
Autoclave or Ethylene Oxide