Learn More
Masterflex™ T-Connecting Union, PVDF
Cole-Parmer™ T-Connecting Union, PVDF is barbed, designed for quick and easy assembly, economical, and suitable for low-pressure applications.
Brand: Masterflex™ 30703-79
Additional Details : Weight : 0.01000kg
Description
- Ideal for low-pressure applications.
- Quick and easy assembly without tools.
- Available in a variety of sizes, and configurations.
- Easily connect fluid paths using flexible tubing.
Specifications
Autoclavable | |
5/8 in. ID tubing | |
10/Pk. | |
Fits 5/8 in. ID tubing |
Barb | |
Low-pressure applications | |
T-connector | |
Autoclave or Ethylene Oxide |