Learn More
Masterflex™ T-Connecting Union, PVDF
Cole-Parmer™ T-Connecting Union, PVDF is barbed, designed for quick and easy assembly, economical, and suitable for low-pressure applications.
530.40 SEK - 678.30 SEK
Specifications
Autoclavable Autoclavable
For Use With (Application) Low-pressure applications
Product Line T-connector
Sterilization Method Autoclave or Ethylene Oxide
Type Barb
View More Specs
Products 3
Product Code Brand Tubing Size Packaging Quantity Price Quantity & Availability  
Product Code Brand Tubing Size Packaging Quantity Price Quantity & Availability  
12363790
View Documents Promotion Details
Masterflex™
30703-78
Fits 1/2 in. ID tubing 10/Pk.
576.30 SEK
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 10-03-2022
Add to basket
12429640
View Documents Promotion Details
Masterflex™
30703-79
Fits 5/8 in. ID tubing 10/Pk.
678.30 SEK
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 21-03-2022
Add to basket
12449640
View Documents Promotion Details
Masterflex™
30703-80
Fits 3/8 in. ID tubing 5/Pk.
530.40 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 18-03-2022
Add to basket
Description
Description
  • Ideal for low-pressure applications.
  • Quick and easy assembly without tools.
  • Available in a variety of sizes, and configurations.
  • Easily connect fluid paths using flexible tubing.
Specifications
Specifications
Autoclavable
T-connector
Barb
Low-pressure applications
Autoclave or Ethylene Oxide