Läs mer
Tocris Bioscience™ t-TUCB
Beskrivning
t-TUCB is a potent soluble epoxide hydrolase (sEH) Inhibitor (IC50 = 0.9 nM). Enhances hepatic eNOS phosphorylation and NO release. Exhibits antihypertensive and cardioprotective actions in cirrhotic rat models. Suppress the activation of HSCs and reduce hepatic fibrosis through inhibition of NF-κB/IκBα signaling. Anti-inflammatory and antinociceptive.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 4-[[trans-4-[[[[4-(Trifluoromethoxy)phenyl]amino]carbonyl]amino]cyclohexyl]oxy]benzoic acid |
| CAS | 948304-40-3 |
| Kvantitet | 5 mg |
| Molekylformel | C21H21F3N2O5 |
| Formel vikt | 438.4 |
| Produkttyp | Inhibitor |
| Renhet | 98% |