missing translation for 'onlineSavingsMsg'
Läs mer

Cocoon Biosciences T4 DNA Ligase

Produktkod. 31722997
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Koncentration:
2 MU/mL
400 kU/mL
Kvantitet:
100 kU
400 kU
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Koncentration Kvantitet
31722997 2 MU/mL 100 kU
31722996 400 kU/mL 100 kU
31722998 400 kU/mL 400 kU
31722999 2 MU/mL 400 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 31722997 Leverantör Cocoon Biosciences Leverantörsnummer SP267126A1

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Enables high-efficiency joining of DNA fragments, supporting cloning, assembly, and repair workflows. Its strong performance improves reaction reliability, boosts ligation success rates, and enhances overall workflow consistency.

  • Catalyzes the formation of phosphodiester bonds between cohesive or blunt ended strands of DNA or repairs single stranded nicks in duplex DNA.
  • Molecular cloning: used for ligation of DNA fragments into plasmid vectors, circularization of DNA molecules, and related applications.
  • DNA repair for joining double strand breaks: seals cohesive or blunt ends.
  • Joining adapters to DNA: ligates DNA fragments with specific adapters or vectors for large scale cloning and sequencing.
  • Source: Engineered Baculovirus carrying T4 DNA Ligase gene from T4 Bacteriophage inoculated in inoculate Trichoplusia ni.
  • Enzyme activity: One unit (U) is defined as the amount of enzyme required to ligate ≥90% of 3 μg of Lambda DNA-Hind III Digest in 20 min at 16°C.
  • Reliable and consistent quality: manufactured under high quality standards, ensuring batch to batch consistency.
  • High concentrated presentation for use in challenging assay conditions.
  • Delivered with 10X Ligation Buffer.
  • Environmentally considerate production prioritizing minimal environmental impact.
  • For research use or further manufacturing; not for diagnostic use or direct administration into humans or animals.

Specifikationer

Koncentration 2 MU/mL
Värd art Enterobacteria phage T4
Färg Transparent
Komponenter Ligation Buffer
Kvalitet RUO
Renhet >95% by SDS-PAGE
pH 7.4
För användning med (applikation) DNA Ligation, DNA repair
Förvaringsbuffert 10 mM Tris-HCl pH 7.4, 50 mM KCl, 1 mM DTT, 0.1 mM EDTA and 50% Glycerol
Förpackning Vial with Lid
Innehåll och lagring -20°C
Uttryckssystem Trichoplusia ni (insect)
Gen-ID (Entrez) 1258680
Gene Alias Polydeoxyribonucleotide synthase [ATP]
Massa 57.5 kDa
Protein Tag His
Frakt skick 2°C to 8°C
Enzym DNA Ligase
Känslig för värmeinaktivering Yes
Kvantitet 100 kU
Produkttyp Enzyme
Forskningskategori Traditional Cloning, Next-generation Sequencing, DNA Repair
Molekylvikt (g/mol) 57455.41
Biologisk aktivitet DNA Ligase
Provtyp Purified Recombinant Protein
Form Liquid
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.