Läs mer
Tabimorelin, TRC
Produktkod. 30008440
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008440 5 mg
30004103 10 mg
30003378 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008440 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT0035005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 193079-69-5
Molekylformel C32 H40 N4 O3
Molekylvikt (g/mol) 528.68
Synonym D-Phenylalaninamide, N-[(2E)-5-amino-5-methyl-1-oxo-2-hexen-1-yl]-N-methyl-3-(2-naphthalenyl)-D-alanyl-N,Nα-dimethyl-, D-Phenylalaninamide, N-[(2E)-5-amino-5-methyl-1-oxo-2-hexenyl]-N-methyl-3-(2-naphthalenyl)-D-alanyl-N,Nα-dimethyl- (9CI), N-[(2E)-5-Amino-5-methyl-1-oxo-2-hexen-1-yl]-N-methyl-3-(2-naphthalenyl)-D-alanyl-N,Nα-dimethyl-D-phenylalaninamide, NN 703, NNC 26-0703, Tabimorelin
IUPAC-namn (E)-5-amino-N,5-dimethyl-N-[(2R)-1-[methyl-[(2R)-1-(methylamino)-1-oxo-3-phenylpropan-2-yl]amino]-3-naphthalen-2-yl-1-oxopropan-2-yl]hex-2-enamide
LEDER CNC(=O)[C@@H](Cc1ccccc1)N(C)C(=O)[C@@H](Cc2ccc3ccccc3c2)N(C)C(=O)\C=C\CC(C)(C)N
Specifikationer
Kemiskt namn eller material Tabimorelin
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C32H40N4O3/c1-32(2,33)19-11-16-29(37)35(4)28(22-24-17-18-25-14-9-10-15-26(25)20-24)31(39)36(5)27(30(38)34-3)21-23-12-7-6-8-13-23/h6-18,20,27-28H,19,21-22,33H2,1-5H3,(H,34,38)/b16-11+/t27-,28-/m1/s1
Formel vikt 528.31
Procent renhet >95%