Läs mer
TACA, TRC
Produktkod. 17922115
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17922115 100 mg
17992465 25 mg
17921365 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17922115 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT004010100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 38090-53-8
Molekylformel C4 H7 N O2
Molekylvikt (g/mol) 101.104
Synonym TACA, trans-4-Amino-2-butenoic Acid, trans-4-Aminocrotonic Acid, (E)-4-Amino-2-butenoic Acid, (2E)-4-Amino-2-butenoic Acid;
IUPAC-namn (E)-4-aminobut-2-enoic acid
LEDER NC\C=C\C(=O)O
Specifikationer
Kemiskt namn eller material 4-trans-Aminocrotonic Acid
Kvantitet 100 mg
InChI formel InChI=1S/C4H7NO2/c5-3-1-2-4(6)7/h1-2H,3,5H2,(H,6,7)/b2-1+
Formel vikt 101.0477