Läs mer
Tacalcitol Monohydrate, TRC
Produktkod. 30428081
Change view
Kvantitet:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30428081 5 mg
30412728 1 mg
30419798 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30428081 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT0040505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 93129-94-3
Molekylformel C27 H44 O3 . H2 O
Molekylvikt (g/mol) 434.65
Synonym 1,3-Cyclohexanediol, 4-methylene-5-[(2E)-2-[(1R,3aS,7aR)-octahydro-1-[(1R,4R)-4-hydroxy-1,5-dimethylhexyl]-7a-methyl-4H-inden-4-ylidene]ethylidene]-, hydrate (1:1), (1R,3S,5Z)-, ,10-Secocholesta-5,7,10(19)-triene-1,3,24-triol, monohydrate, (1alpha,3beta,5Z,7E,24R)- (9CI), 1alpha,24(R)-Dihydroxy-vitamin D3 monohydrate, 1alpha,24(R)-Dihydroxycholecalciferol monohydrate, Tacalcitol monohydrate
IUPAC-namn (1R,3S,5Z)-5-[(2E)-2-[(1R,3aS,7aR)-1-[(2R,5R)-5-hydroxy-6-methylheptan-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexane-1,3-diol;hydrate
LEDER O.CC(C)[C@H](O)CC[C@@H](C)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@@H](O)C[C@H](O)C3=C)\CCC[C@]12C
Specifikationer
Kemiskt namn eller material Tacalcitol Monohydrate
Kvantitet 5 mg
InChI formel InChI=1S/C27H44O3.H2O/c1-17(2)25(29)13-8-18(3)23-11-12-24-20(7-6-14-27(23,24)5)9-10-21-15-22(28)16-26(30)19(21)4;/h9-10,17-18,22-26,28-30H,4,6-8,11-16H2,1-3,5H3;1H2/b20-9+,21-10-;/t18-,22-,23-,24+,25-,26+,27-;/m1./s1
Formel vikt 434.3396