missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACO1/CCDC44 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21147-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
TACO1 is a mitochondrial translational activator required for efficient translation of cytochrome c oxidase (Weraarpachai et al., 2009), a component of complex IV of the mitochondrial respiratory complex.Specifications
TACO1/CCDC44 | |
Polyclonal | |
Unconjugated | |
TACO1 | |
CCDC44, TACO1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
360645, 51204, 70207 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B2RYT9, Q8K0Z7, Q9BSH4 | |
Taco1 | |
TACO1/CCDC44 Fusion Protein Ag15688 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |