Läs mer
TACS, TRC
Produktkod. 30418640
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30418640 10 mg
30424708 100 mg
30428235 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30418640 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT00420010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 111298-82-9
Molekylformel C14H15N3O5S3
Molekylvikt (g/mol) 401.48
Synonym (6R,7R)-7-Amino-3-[[[5-(carboxymethyl)-4-methyl-2-thiazolyl]thio]methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, (6R-trans)-7-Amino-3-[[[5-(carboxymethyl)-4-methyl-2-thiazolyl]thio]methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid
IUPAC-namn (6R,7R)-7-amino-3-[[5-(carboxymethyl)-4-methyl-1,3-thiazol-2-yl]sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
LEDER Cc1nc(SCC2=C(N3[C@H](SC2)[C@H](N)C3=O)C(=O)O)sc1CC(=O)O
Specifikationer
Kemiskt namn eller material TACS
Kvantitet 10 mg
InChI formel InChI=1S/C14H15N3O5S3/c1-5-7(2-8(18)19)25-14(16-5)24-4-6-3-23-12-9(15)11(20)17(12)10(6)13(21)22/h9,12H,2-4,15H2,1H3,(H,18,19)(H,21,22)/t9-,12-/m1/s1
Formel vikt 401.0174