missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TADA2L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567425
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: MDRLGPFSNDP SDKPPCRGCS SYLMEPYIKC AECGPPPFFL CLQCFTRGFE YKKHQSDHTY EIMTSD Highest antigen sequence identity to the following orthologs - mouse 97%, rat 99%.
TADA2L Polyclonal antibody specifically detects TADA2L in Human samples. It is validated for Immunocytochemistry.Specifications
TADA2L | |
Polyclonal | |
Unconjugated | |
TADA2A | |
ADA2; ADA2A; ADA2-like protein; AV319371; D030022J10Rik; hADA2; KL04P; TADA2A; TADA2L; transcriptional adapter 2-alpha; Transcriptional adapter 2-like; transcriptional adaptor 2 alpha; transcriptional adaptor 2A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6871 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75478 | |
TADA2A | |
Recombinant Human TADA2L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only