Läs mer
Tadalafil Ketolactam (>90%), TRC
Produktkod. 30423185
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30423185 10 mg
30422339 25 mg
30412169 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30423185 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT00453510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1346605-38-6
Molekylformel C22H19N3O6
Molekylvikt (g/mol) 421.4
Synonym (6R,14aR)-rel-6-(1,3-Benzodioxol-5-yl)-2,3,14,14a-tetrahydro-2-methylpyrazino[1,2-d][1,4]benzodiazonine-1,4,7,13(6H,8H)-tetrone, Tadalafil EP Impurity H
IUPAC-namn (6R,14aR)-6-(benzo[d][1,3]dioxol-5-yl)-2-methyl-2,3,14,14a-tetrahydrobenzo[h]pyrazino[1,2-d][1,4]diazonine-1,4,7,13(6H,8H)-tetraone
LEDER O=C1N(C)CC(N([C@]1([H])CC(C(C=CC=C2)=C2N3)=O)[C@@](C4=CC=C(OCO5)C5=C4)([H])C3=O)=O
Specifikationer
Kemiskt namn eller material Tadalafil Ketolactam
Kvantitet 10 mg
InChI formel InChI=1S/C22H19N3O6/c1-24-10-19(27)25-15(22(24)29)9-16(26)13-4-2-3-5-14(13)23-21(28)20(25)12-6-7-17-18(8-12)31-11-30-17/h2-8,15,20H,9-11H2,1H3,(H,23,28)/t15-,20-/m1/s1
Formel vikt 421.13