Läs mer
Tocris Bioscience™ Tadalafil
GREENER_CHOICE
Produktkod. 30000939
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30000939 10 mg
30000921 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30000939 Leverantör Tocris Bioscience™ Leverantörsnummer 6311/10
Potent and highly selective PDE5 inhibitor; orally bioavailable
Potent and highly selective PDE5 inhibitor (IC50 = 5 nM). Exhibits > 2000-fold selectivity for PDE5 over PDEs 1,2,3 and 4 and 1000-fold selectivity over PDE6. Reduces blood pressure in hypertensive rats. Also delays tumor growth in a mouse model via inhibition of MDSC-mediated immunosuppression. Orally bioavailable.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 171596-29-5
Molekylformel C22H19N3O4
Specifikationer
Kemiskt namn eller material (6R,12aR)-6-(1,3-Benzodioxol-5-yl)-2,3,6,7,12,12a-hexahydro-2-methylpyrazino[1',2':1,6]pyrido[3,4-b]
Kvantitet 10 mg
Löslighet Soluble to 100mM in DMSO
Formel vikt 389.4
Fysisk form Solid
Renhet >99%
Molekylvikt (g/mol) 389.411