Learn More
Thermo Scientific Chemicals TAE (10X), TRIS + acetate + EDTA
TAE (10X), TRIS + acetate + EDTA is a buffer composed of Tris, acetate, and EDTA components and is used in electrophoresis of nucleic acids in agarose and polyacrylamide gels.
629.56 SEK - 1680.39 SEK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15425859
|
Thermo Scientific Chemicals
J63677.K2 |
1 L |
629.56 SEK
1 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15435859
|
Thermo Scientific Chemicals
J63677.K3 |
2 L |
931.32 SEK
2 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15445859
|
Thermo Scientific Chemicals
J63677.K7 |
4 L |
1680.39 SEK
4 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TAE is used for DNA agarose gel electrophoresis but is also used for non-denaturing RNA agarose gel electrophoresis, used at various concentrations to study the mobility of DNA in solution.