Learn More
Thermo Scientific Chemicals TAE (10X), TRIS + acetate + EDTA
TAE (10X), TRIS + acetate + EDTA is a buffer composed of Tris, acetate, and EDTA components and is used in electrophoresis of nucleic acids in agarose and polyacrylamide gels.
629.56 SEK - 1680.39 SEK
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15425859
SDS View Documents Promotion Details
Thermo Scientific Chemicals
J63677.K2
1 L
629.56 SEK
1 L
View alternative products
Save Up to 
»
In Stock
to see stock.
15435859
SDS View Documents Promotion Details
Thermo Scientific Chemicals
J63677.K3
2 L
931.32 SEK
2 L
View alternative products
Save Up to 
»
In Stock
to see stock.
15445859
SDS View Documents Promotion Details
Thermo Scientific Chemicals
J63677.K7
4 L
1680.39 SEK
4 L
View alternative products
Save Up to 
»
In Stock
to see stock.
Description
Description
TAE is used for DNA agarose gel electrophoresis but is also used for non-denaturing RNA agarose gel electrophoresis, used at various concentrations to study the mobility of DNA in solution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• TAE (10X), TRIS + acetate + EDTA allows nucleic acid separation
Application
• This buffer has been used to prepare agarose and polyacrylamide gels for electrophoresis of nucleic acids
• It is used as a running and gel preparation buffer, and it is also used for non-denaturing RNA agarose gel electrophoresis