missing translation for 'onlineSavingsMsg'
Learn More

TAF10 Monoclonal Antibody (23TA-1H8), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA3069

Additional Details : Weight : 0.01000kg

Product Code. 15782526

  • 4210.00 SEK / 50µL
Estimated Shipment: 30-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MA3-069 detects TAF10 from human, rat, and non-human primate samples. MA3-069 does not detect the TAF10 in mouse samples MA3-069 has been successfully used in Western blot, immunofluorescence and flow cytometry applications. The MA3-069 immunogen is the synthetic peptide coupled to ovalalbumin aa 1 to aa 20.

TAF10 Monoclonal antibody specifically detects TAF10 in Human, Non-human Primate, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.
Specifications

Specifications

TAF10
Monoclonal
Conc. Not Determined
ascites with 0.05% sodium azide
Q12962
TAF10
Synthetic peptide coupled to ovalalbumin aa 1 to 20
RUO
293345, 6881
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry, Immunocytochemistry, Western Blot
23TA-1H8
Unconjugated
TAF10
30kDa; AU041226; im:6899389; mTAFII30; si:dkey-72l14.8; STAF28; TAF(II)30; TAF10; TAF10 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF10 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 30 kDa; TAF10 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 30kDa; TAF2A; Taf2h; TAFII30; TAFII-30; TATA box binding protein (TBP)-associated factor, RNA polymerase II, H, 30 kDa; TATA box binding protein (TBP)-associated factor, RNA polymerase II, H, 30kD; TATA box binding protein associated factor 10; TATA-box binding protein associated factor 10; TBP-related factor 10; transcription initiation factor TFIID 30 kD subunit; transcription initiation factor TFIID 30 kDa subunit; Transcription initiation factor TFIID subunit 10
Mouse
50 μL
Primary
Human, Non-human Primate, Rat
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers